Hydrogeenbromide 47%, NORMATOM®, Ultrapure for trace metal analysis

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Hydrogen bromide

85032.270EA 1150 EUR
85032.270
Hydrogeenbromide 47%, NORMATOM®, Ultrapure for trace metal analysis
Hydrogeenbromide
Formule: HBr
MW: 80.91 g/mol
Kookpunt: -66.7
Smeltpunt: -86
Dichtheid: 1.47 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011323
CAS nummer: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8 II
Merck Index: 13,04799

Order Now

Specification Test Results

Assay 44-49%
Ag (Silver) 100 ppt
Al (Aluminium) 100 ppt
Au (Gold) 100 ppt
B (Boron) Information only
Ba (Barium) 100 ppt
Be (Beryllium) 100 ppt
Bi (Bismuth) 100 ppt
Ca (Calcium) 100 ppt
Cd (Cadmium) 100 ppt
Ce (Cerium) 10 ppt
Co (Cobalt) 100 ppt
Cr (Chromium) 100 ppt
Cs (Cesium) 10 ppt
Cu (Copper) 100 ppt
Dy (Dysprosium) 10 ppt
Er (Erbium) 10 ppt
Eu (Europium) 10 ppt
Fe (Iron) 100 ppt
Ga (Gallium) 100 ppt
Gd (Gadolinium) 10 ppt
Hf (Hafnium) 10 ppt
Ho (Holmium) 10 ppt
In (Indium) 10 ppt
K (Potassium) 100 ppt
La (Lanthanum) 10 ppt
Li (Lithium) 100 ppt
Lu (Lutetium) 10 ppt
Mg (Magnesium) 100 ppt
Mn (Manganese) 100 ppt
Mo (Molybdenum) 100 ppt
Na (Sodium) 100 ppt
Nb (Niobium) 10 ppt
Nd (Neodymium) 10 ppt
Ni (Nickel) 100 ppt
Pb (Lead) 1 ppt
Pd (Palladium) 100 ppt
Pr (Praseodymium) 10 ppt
Pt (Platinum) 100 ppt
Rb (Rubidium) 100 ppt
Re (Rhenium) 100 ppt
Rh (Rhodium) 100 ppt
Ru (Ruthenium) 100 ppt
Sb (Antimony) Information only
Sc (Scandium) 100 ppt
Sm (Samarium) 10 ppt
Sn (Tin) 100 ppt
Sr (Strontium) 100 ppt
Ta (Tantalum) Information only
Tb (Terbium) 10 ppt
Te (Tellurium) 10 ppt
Th (Thorium) 1 ppt
Ti (Titanium) 100 ppt
Tl (Thallium) 10 ppt
Tm (Thulium) 10 ppt
U (Uranium) 1 ppt
V (Vanadium) 100 ppt
W (Tungsten) 100 ppt
Y (Yttrium) 10 ppt
Yb (Ytterbium) 10 ppt
Zn (Zinc) 100 ppt
Zr (Zirconium) 10 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR