Mierenzuur ≥99%, HiPerSolv CHROMANORM® for LC-MS

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Packaged under nitrogen.

85048.001EA 120 EUR
85048.001 85048.010 85048.051
Mierenzuur ≥99%, HiPerSolv CHROMANORM® for LC-MS
Mierenzuur

LC-MS additives are selective chemicals that are commonly added to the mobile phase in LC-MS. The objective is usually to improve the signal quality, but they can also be used to suppress unwanted signals or selectively enhance the signal of specific compounds in a mixture


  • High quality and LC-MS application tested
  • Improved ionisation and resolution

Formule: HCO₂H
Kookpunt: 100…101 °C (1013 hPa)
Smeltpunt: 8,2…8,4 °C
Dichtheid: 1,22 g/cm³ (25 °C)
Vlampunt: 48 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003297
CAS nummer: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 (3),II
Bereik: 01-2119491174-37
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min 99 %
Evaporation residue Max 0.0001 %
Ca (Calcium) (0.1 %) Max 0.2 ppm
K (Potassium) (0.1 %) Max 0.2 ppm
Mg (Magnesium) (0.1 %) Max 0.2 ppm
Na (Sodium) (0.1 %) Max 0.5 ppm
Transmittance (225 nm)(0.1 %) Min 20 %
Transmittance (235 nm)(0.1 %) Min 50 %
Transmittance (240 nm)(0.1 %) Min 80 %
Transmittance (250 nm)(0.1 %) Min 95 %
Transmittance (260 nm)(0.1 %) Min 99 %
Suitability for LC-MS (0.1 %) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR