Zilvernitraat ≥99.5% (by argentometric titration), Puriss. p.a., Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

85228-250G 85228-50G
85228-250GEA 1040 EUR
85228-250G 85228-50G
Zilvernitraat ≥99.5% (by argentometric titration), Puriss. p.a., Fluka™
Zilvernitraat
Formule: AgNO₃
Kookpunt: Dec. (on heating)
Smeltpunt: 212 °C
Dichtheid: 4,350 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003414
CAS nummer: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
Bereik: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5 - 100.5 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 1 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 2 ppm
Iron (Fe) Max. 2 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 1 ppm
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR