Ammonium thiocyanate in aqueous solution USP Test Solution (TS)
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Synoniemen: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate
85331.180EA 43 EUR
85331.180 85331.260
Ammonium thiocyanate in aqueous solution USP Test Solution (TS)
Ammonium thiocyanate
Ready to use test solution (TS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Formule: NH₄SCN
Smeltpunt: 150 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011428
CAS nummer: 1762-95-4
EINECS: 0
Merck Index: 13,00564