Kopersulfaat 0.5 mol/l USP Test Solution (TS)

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

85343.180EA 42.8 EUR
85343.180 85343.260
Kopersulfaat 0.5 mol/l USP Test Solution (TS)
Koper(II)sulfaat

Ready to use test solution (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formule: CuSO₄
Smeltpunt: 200 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010981
CAS nummer: 7758-98-7
EINECS: 0
UN: 3082
ADR: 9,III
Merck Index: 12,02722

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR