N,O-Bis(trimethylsilyl)acetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSA+TMCS+TMSI) silylating mixture II according to Horning, LiChropur™ for GC derivatisation, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ100

85432-10ML 85432-1ML 85432-10X1ML
85432-10MLEA 163 EUR
85432-10ML 85432-1ML 85432-10X1ML
N,O-Bis(trimethylsilyl)acetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSA+TMCS+TMSI) silylating mixture II according to Horning, LiChropur™ for GC derivatisation, Supelco®
N,O-Bis(trimethylsilyl)acetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSA+TMCS+TMSI) mixture
Vlampunt: 4 °C
UN: 2924
ADR: 3 (8) II

Order Now

Specification Test Results

Appearance (Colour) Faint Yellow to Yellow
Appearance (Form) Liquid
Refractive index N20/D 1,434 - 1,438
Proton NMR Spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR