Zoutzuur 1 mol/l (1 N) voor biofarmaceutische productie

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

85447.320EA 37.5 EUR
85447.320 85447.460
Zoutzuur 1 mol/l (1 N) voor biofarmaceutische productie
Zoutzuur
Formule: HCl
Dichtheid: 1,014 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Appearance Passes test
Free bromine or chlorine Passes test
Identification Passes test
Colouration Max. 10 APHA
Evaporation residue Max. 100 ppm
Free chlorine Max. 1 ppm
Heavy metals (as Pb) Max. 0.5 ppm
Ignition residue (SO4) Max. 80 ppm
SO3 (Sulphite) Max. 10 ppm
SO4 (Sulphate) Max. 5 ppm
As (Arsenic) Max. 0.1 ppm
Residual metal catalysts Unlikely to be present
Residual solvents (ICH Q3C) Excl.by manufact. process
Endotoxin Max. 0,5 EU/ml
Filtered at 0,2 µm Confirmed
Titer 0.95 - 1.05 mol/l

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR