VWR®, Coulometric reagents for water determination by the Karl Fischer method
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
85470.290EA 228 EUR
85470.290 85471.130 85472.260 85473.260 85474.260 85475.260 85476.260 85477.260 85478.260 85479.130
VWR®, Coulometric reagents for water determination by the Karl Fischer method
Karl Fischer titranten en reagentia
Both anolyte and catholyte solutions are with improved formulation to increase the speed and accuracy of titration when determining water content at the microgram level.
  • Reach endpoint quickly and consistently
  • Long shelf life
  • Stable endpoint
Order Now
Specificaties
Environmentally Preferable