VWR®, Poederpakketten voor fosfaatbepaling, reagentia voor fotometers
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
85621.610EA 32.4 EUR
85621.610 85621.680 85622.610 85623.610 85624.610
VWR®, Poederpakketten voor fosfaatbepaling, reagentia voor fotometers
Milieutestkits
VWR® powder pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary. The pillows, individually sealed in aluminium foil packs, are unaffected by ambient conditions and contain pre-measured powder reagents for accuracy, convenience and great solubility.
  • Accurate
  • Fast and convenient by easier dissolution than tablets and long shelf life
  • No calibration necessary
  • Beveled edge prevents reagent from getting stuck in corners
PP = Powder Pack
RS = Powder Pack reagentia-set