Tetrabutylammonium hydroxide 20% in aqueous solution, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

85740.180EA 43.6 EUR
85740.180 85740.260
Tetrabutylammonium hydroxide 20% in aqueous solution, GPR RECTAPUR®
Tetrabutylammonium hydroxide
Formule: C₁₆H₃₇NO
MDL Nummer: MFCD00009425
CAS nummer: 2052-49-5
UN: 3267
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR