VWR® ARISTAR®, Cyanide 1000 mg/L Standard for ion chromatography in 0.1% KOH, Certified Reference Material

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

85784.260EA 99.2 EUR
85784.260
VWR® ARISTAR®, Cyanide 1000 mg/L Standard for ion chromatography in 0.1% KOH, Certified Reference Material
Standaarden Anionische standaarden Cyanide standaarden

This certified reference material is produced using a high purity starting material, acid from sub-boiling and 18 MOhm deionized water and filtered through a 0.2 micron filter.


Certified value and uncertainty (mg/l): 1000.0 ± 5.0


Certificaten: CRM ISO 17034 and ISO 17025. Traceable to NIST.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR