Ethanol absolute ≥99.8%, PESTINORM® for pesticide residue analysis

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Ethyl alcohol

85933.290EA 48.9 EUR
85933.290 85933.320
Ethanol absolute ≥99.8%, PESTINORM® for pesticide residue analysis
Ethanol
Formule: C₂H₅OH
Kookpunt: 78,3 °C (1013 hPa)
Smeltpunt: –117 °C
MDL Nummer: MFCD00003568
CAS nummer: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless liquid
Assay (GC, on anhydrous basis) min. 99.8 %
Identity (IR) Comply
Residue after evaporation max. 0.0003 % w/w
Water (KF) max. 0.05 % w/w
GC/ECD any Pesticide (as Lindane) max. 5 pg/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR