Zoutzuur 0.1 mol/l (0.1 N) 2-propanol acc. to DIN 51558 part 1 , AVS TITRINORM

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Hydrochloric acid solution, Muriatic acid

86170.290EA 85.5 EUR
86170.290
Zoutzuur 0.1 mol/l (0.1 N) 2-propanol acc. to DIN 51558 part 1 , AVS TITRINORM
Zoutzuur
Formule: HCl
MW: 36.46 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
UN: 2920
ADR: 8 I

Order Now

Specification Test Results

Titer (20°C) (result of analysis at point of filling) 0.9980 – 1.0020


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR