VWR®, Holmium standard solution, 1,000 mg/L Ho in dilute nitric acid AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
86683.180EA 29.96 EUR
86683.180 86683.260
VWR®, Holmium standard solution, 1,000 mg/L Ho in dilute nitric acid AVS TITRINORM standard for AAS
Standaarden Kationische standaarden Holmium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%