VWR® AVS TITRINORM, Lead Standard Solution, 1000 mg/L Pb in dil. Nitric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
86699.260EA 36.1 EUR
86699.260 86699.180
VWR® AVS TITRINORM, Lead Standard Solution, 1000 mg/L Pb in dil. Nitric Acid, Standard for AAS
Standaarden Kationische standaarden Lood standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%