Diisopropylamine ≥99.0%, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

87031.290EA 67.3 EUR
87031.290 87031.320 87031.460
Diisopropylamine ≥99.0%, GPR RECTAPUR®
Diisopropylamine
Formule: C₆H₁₅N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008862
CAS nummer: 108-18-9
UN: 1158
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellowish liquid
Assay (GC)
Relative density (20/4°C)
Water (K. F.)
Identity (IR) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR