n-Nonane ≥99%, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

87100.180EA 42.3 EUR
87100.180 87100.230 87100.290 87100.320
n-Nonane ≥99%, GPR RECTAPUR®
n-Nonane
Formule: H₃C(CH₂)₇CH₃
Vlampunt: 31 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009574
CAS nummer: 111-84-2
EINECS: 203-913-4
UN: 1920
ADR: 3,III

Order Now

Specification Test Results

Assay ≥ 99,0 %
Density (20/4) 0,717 - 0,719


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR