Dimethyl sulphoxide-[D₆] (99.8% D) for NMR spectroscopy

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6

Filtered through a 0.2 µm filter, packaged under nitrogen.

87154.0006EA 44.4 EUR
87154.0006 87154.0007 87154.0025 87154.0010 87154.0100 87154.0011
Dimethyl sulphoxide-[D₆] (99.8% D) for NMR spectroscopy
Dimethyl sulphoxide-[D₆]
Formule: D₃CS(O)CD₃
Kookpunt: 189 °C (5 mmHg)
Dichtheid: 1,19 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002090
CAS nummer: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Isotopic enrichment (FT NMR 400 MHz)(D) Min. 99.80 %
Water Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR