Bromothymol blue sodium salt 0,05% in ethanol 20% (v/v) Reag. Ph. Eur. 1012901

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

87701.180EA 45.2 EUR
87701.180
Bromothymol blue sodium salt 0,05% in ethanol 20% (v/v) Reag. Ph. Eur. 1012901
Bromothymol blue sodium salt

Bestelinformatie: Normally manufactured to order. Please check with customer services.

Formule: C₂₇H₂₇Br₂NaO₅S
MDL Nummer: MFCD00077263
CAS nummer: 34722-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR