Copper tetramine complex in aqueous solution (copper(II) hydroxide 10% in ammonia solution 20%) Reag. Ph. Eur. 1022600

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

87809.180EA 252 EUR
87809.180
Copper tetramine complex in aqueous solution (copper(II) hydroxide 10% in ammonia solution 20%) Reag. Ph. Eur. 1022600
Copper tetramine complex in aqueous solution (copper(II) hydroxide 10% in ammonia solution 20%)

Bestelinformatie: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formule: Cu(OH)₂
CAS nummer: 20427-59-2
EINECS: 243-815-9
UN: 3266
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR