Basic Violet 3 solution Reag. Ph. Eur. 1022901

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: CI 42555

87811.180EA 24.2 EUR
87811.180
Basic Violet 3 solution Reag. Ph. Eur. 1022901
Basic Violet 3

Bestelinformatie: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formule: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
MDL Nummer: MFCD00011750
CAS nummer: 548-62-9
UN: 2789
ADR: 8 (3) II
Merck Index: 13,04406

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR