Fenolrood natriumzout 0,1% in ethanol (20%) Reag. Ph. Eur. 1063601

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

87888.180EA 45.2 EUR
87888.180
Fenolrood natriumzout 0,1% in ethanol (20%) Reag. Ph. Eur. 1063601
Fenolrood natriumzout

Bestelinformatie: Normally manufactured to order. Please check with customer services.

Formule: C₁₉H₁₃NaO₅S
MDL Nummer: MFCD00066901
CAS nummer: 34487-61-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR