Salpeterzuur 65 - 70% ≥99.999% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

87920.AE 87920.AP
87920.AEEA 69.7 EUR
87920.AE 87920.AP
Salpeterzuur 65 - 70% ≥99.999% (metals basis)
Salpeterzuur

Purity: 99.999% CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Density: 1.41 Appearance: Colourless to pale yellow Odour: Pungent Boiling Point: 121° Solubility: Very soluble in water and ether. Decomposes in alcohol Merck Reference: 14,6577 UN No.: 2031 MDL No.: MFCD00011349


In the manufacture of fertilizers and explosives, in organic synthesis, in metallurgy, in photoengraving, in etching steel, in rubber chemicals, and in reprocessing nuclear fuels

Formule: HNO₃
Kookpunt: 121 °C
Smeltpunt: –42 °C
Dichtheid: 1,41
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011349
CAS nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 14,6577

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR