Potassium thiocyanate 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1071801

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Potassium rhodanide

87922.290EA 132 EUR
87922.290
Potassium thiocyanate 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1071801
Potassium thiocyanate

Bestelinformatie: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formule: KSCN
MW: 97.18 g/mol
Kookpunt: 500
Smeltpunt: 175
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011413
CAS nummer: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR