Zilvernitraat 0.25 mol/l (0.25 N) in aqueous solution Reag. Ph. Eur. 1078301

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

87924.290EA 198 EUR
87924.290
Zilvernitraat 0.25 mol/l (0.25 N) in aqueous solution Reag. Ph. Eur. 1078301
Zilvernitraat

Bestelinformatie: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formule: AgNO₃
Dichtheid: 1,033 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003414
CAS nummer: 7761-88-8
EINECS: 231-853-9
UN: 3264
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Performance test Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR