Natriumchloride 20% aqueous solution Reag. Ph. Eur. 1079502

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Saline, NaCl, Sea salt, Halite

87931.290EA 132 EUR
87931.290
Natriumchloride 20% aqueous solution Reag. Ph. Eur. 1079502
Natriumchloride

Bestelinformatie: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formule: NaCl
MW: 58.44 g/mol
Kookpunt: 1413
Smeltpunt: 801
Dichtheid: 1…1.2 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003477
CAS nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Limited Time Sale

Select items below are currently on sale

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR