Natriumhydroxide 2 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1081402

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Caustic soda

87934.290EA 71.8 EUR
87934.290
Natriumhydroxide 2 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1081402
Natriumhydroxide

Bestelinformatie: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formule: NaOH
Kookpunt: 109 °C (1013 hPa)
Dichtheid: <1,11 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR