Thallium(III) nitrate trihydrate ≥99.5% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Thallium trinitrate trihydrate

87999.06 87999.14 87999.22
87999.06EA 64.1 EUR
87999.06 87999.14 87999.22
Thallium(III) nitrate trihydrate ≥99.5% (metals basis)
Thallium(III) nitrate trihydrate

Purity: 99.5% CAS No.: 13453-38-8 Molecular Formula: Tl(NO3)3.3H2O Formula Weight: 444.43 (390.38anhy) Physical Form: Crystalline Sensitivity: Hygroscopic UN No.: 2727 MDL No.: MFCD00149874

Formule: N₃O₉Tl·3H₂O
MW: 444.44 g/mol
Smeltpunt: 102…105 °C
Dichtheid: 3.31…3.37 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149874
CAS nummer: 13453-38-8
EINECS: 237-325-4
UN: 2727
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR