Boron standard solution, 1000 mg/L B in water (from B(OH₃)), standard for AAS, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

88055.AE 88055.AP
88055.AEEA 40.7 EUR
88055.AE 88055.AP
Boron standard solution, 1000 mg/L B in water (from B(OH₃)), standard for AAS, Specpure®
Standaarden Kationische standaarden Borium standaarden

Molecular Formula: H₃BO₃ in H₂O
Physical Form: Liquid
MDL No.: MFCD00134034


Certificaten: Traceable to NIST.

Order Now

Specificaties

Environmentally Preferable
MDL No. MFCD00134034

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR