Gadolinium standard solution, 1000 mg/L Gd in 5% nitric acid (de Gd?O?), standard for AAS, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

88065.AE 88065.AP
88065.AEEA 44.8 EUR
88065.AE 88065.AP
Gadolinium standard solution, 1000 mg/L Gd in 5% nitric acid (de Gd?O?), standard for AAS, Specpure®
Standaarden Kationische standaarden Gadolinium standaarden

Molecular Formula: Gd₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011022


Certificaten: Traceable to NIST

Order Now

Specificaties

Environmentally Preferable
MDL No. MFCD00011022

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR