Potassium standard solution, 1000 mg/L K in 5% nitric acid (from KNO₃), standard for AAS, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

88087.AE 88087.AP
88087.AEEA 38.9 EUR
88087.AE 88087.AP
Potassium standard solution, 1000 mg/L K in 5% nitric acid (from KNO₃), standard for AAS, Specpure®
Standaarden Kationische standaarden Kalium standaarden

Molecular Formula: KNO₃ 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00133776


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR