Lead(II) telluride ≥99.999% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

88285.14 88285.22
88285.14EA 176 EUR
88285.14 88285.22
Lead(II) telluride ≥99.999% (metals basis)
Lead(II) telluride

Purity: 99.999% CAS No.: 1314-91-6 Molecular Formula: PbTe Formula Weight: 334.79 Physical Form: 15mm & Down Pieces Density: 8.164 Melting Point: 917° Merck Reference: 14,5422 UN No.: 3284 MDL No.: MFCD00016274

Formule: PbTe
Smeltpunt: 917 °C
Dichtheid: 8,164
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016274
CAS nummer: 1314-91-6
EINECS: 215-247-1
UN: 3077
ADR: 9,III
Merck Index: 14,5422

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR