Dipotassium hydrogen orthophosphate trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate

88363.230EA 64.5 EUR
88363.230 88363.290 88363.360 88363.460
Dipotassium hydrogen orthophosphate trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Dipotassium hydrogen orthophosphate trihydrate
Formule: K₂HPO₄·3H₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149926
CAS nummer: 16788-57-1

Order Now

Specification Test Results

Assay (acidimetric) ≥ 99,0 %
Solution in water Passes test
pH (25°C; 5 %) 9,1 - 9,3
Heavy metals (as Pb) ≤ 10 ppm
Total N (Nitrogen) ≤ 10 ppm
Cl (Chloride) ≤ 10 ppm
SO4 (Sulphate) ≤ 50 ppm
As (Arsenic) ≤ 0,5 ppm
Cu (Copper) ≤ 3 ppm
Fe (Iron) ≤ 5 ppm
Na (Sodium) ≤ 0,1 %
Pb (Lead) ≤ 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR