Iron(II) gluconate hydrate ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

88726.36 88726.A3
88726.36EA 56.5 EUR
88726.36 88726.A3
Iron(II) gluconate hydrate ≥97%
Iron(II) gluconate hydrate

Purity: 97% CAS No.: 22830-45-1 Molecular Formula: Fe[HOCH2(CHOH)4CO2]2.xH2O (x≈2) Formula Weight: 446.19(anhy) Physical Form: Granular Melting Point: ca 140° MDL No.: MFCD00150872

Formule: C₁₂H₂₄FeO₁₅
Smeltpunt: ∼ 140 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150872
CAS nummer: 22830-45-1
EINECS: 206-076-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR