VWR®, Conductivity standards, Certified Reference Materials (CRMs) for primary calibration of conductivity meters
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
89001.230EA 113 EUR
89001.230 89002.270 89003.270 89005.270 89006.270 89007.270 89008.270 89009.270 89010.270 89011.270 89012.270 89013.270 89014.270 89015.270 89019.270 89020.270 89021.270 89023.270 89026.270 89027.270 89028.270 89029.270 89030.270 89031.270
VWR®, Conductivity standards, Certified Reference Materials (CRMs) for primary calibration of conductivity meters
Standaarden Geleidbaarheidsstandaarden
Avantor offers a comprehensive range of conductivity standards as Certified Reference Materials with the primary use calibrating of conductivity meters.
  • Accredited acc. ISO 17025 / ISO 17034
  • Traceable to NIST
  • Certificate of Analysis including temperature correction chart
Intended uses:
- Calibration, qualification and monitoring of instruments for conductivity measurement
- Validation of analytical methods
- Preparation of working reference samples
- Detection limit and linearity studies
These standards are produced by dissolving potassium chloride (KCl) in 18 MOhm of deionised water, except where indicated, then filtered through a 0,22 µm filter in equilibrium with atmospheric carbon dioxide. The certified value and expanded uncertainty are obtained using conductivity meter calibration according to ISO/IEC 17025. Homogeneity and stability studies are performed according to the requirement of ISO 17034 and ISO Guide 35. All conductivity standards are traceable to SI (NIST).