Potassium hexafluorosilicate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Potassium fluorosilicate

89023.A1 89023.A7
89023.A1EA 90 EUR
89023.A1 89023.A7
Potassium hexafluorosilicate ≥98%
Potassium hexafluorosilicate

Purity: 98% CAS No.: 16871-90-2 Molecular Formula: K2SiF6 Formula Weight: 220.28 Physical Form: Powder Density: 2.27 pH: 3.4 Melting Point: dec. Solubility: Slightly soluble in cold water. Hydrolyzes in hot water. Insoluble in alcohol Merck Reference: 14,7638 UN No.: 2655 MDL No.: MFCD00040666

Formule: F₆K₂Si
MW: 220.27 g/mol
Smeltpunt: Dec.
Dichtheid: 2.27
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00040666
CAS nummer: 16871-90-2
EINECS: 240-896-2
UN: 2655
ADR: 6.1 III
Merck Index: 12,07803

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR