Kiezelgel, orange/green, beads, GPR RECTAPUR® 2 - 5 mm, with indicator

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

89054.292EA 29.7 EUR
89054.292 89054.360
Kiezelgel, orange/green, beads, GPR RECTAPUR® 2 - 5 mm, with indicator
Kiezelgel

Our silica gel orange/green is used where products need to be protected from humidity during storage and transportation. The colour changes from orange to dark green when saturated.


  • Mixture of various indicators and free of dangerous materials
  • Colour change from orange to dark green
  • Regeneration at 120 °C for 4 h

Formule: SiO₂
Kookpunt: 2230 °C (1013 hPa)
Smeltpunt: > 1600 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011232
CAS nummer: 7631-86-9

Order Now

Specification Test Results

Absorption capacity (H₂O; 25 °C; 50% RH) ≥20,0%
Residual humidity (150 °C) ≤3,0%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR