Natriumnitriet 0.1 N, AVS TITRINORM Volumetric

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Diazotizing salts

89295.320EA 24.8 EUR
89295.320
Natriumnitriet 0.1 N, AVS TITRINORM Volumetric
Natriumnitriet
Formule: NaNO₂
MW: 69 g/mol
Kookpunt: 320
Smeltpunt: 284
MDL Nummer: MFCD00011118
CAS nummer: 7632-00-0
Merck Index: 13,08721

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR