Copper(II) iodate, watervrij ≥95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

89459.04 89459.09 89459.18
89459.04EA 80.1 EUR
89459.04 89459.09 89459.18
Copper(II) iodate, watervrij ≥95%
Copper(II) iodate

Purity: 95% CAS No.: 13454-89-2 Molecular Formula: Cu(IO3)2 Formula Weight: 413.35 Physical Form: Powder Density: 5.241 Melting Point: dec. UN No.: 1479 MDL No.: MFCD00016055

Formule: CuI₂O₆
Smeltpunt: Dec.
Dichtheid: 5,241
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016055
CAS nummer: 13454-89-2
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR