Silver(I) sulphide ≥99.9% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Disilver sulphide

89473.06 89473.14
89473.06EA 80.5 EUR
89473.06 89473.14
Silver(I) sulphide ≥99.9% (metals basis)
Silver(I) sulphide

Purity: 99.9+% CAS No.: 21548-73-2 Molecular Formula: Ag2S Formula Weight: 247.80 Physical Form: Crystalline powder Density: 7.317 Appearance: Dark grey to black Odour: Odourless Melting Point: 825° Merck Reference: 14,8530 UN No.: 3077 MDL No.: MFCD00003406

Formule: Ag₂S
MW: 247.8 g/mol
Smeltpunt: 825 °C
Dichtheid: 7.317
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003406
CAS nummer: 21548-73-2
EINECS: 244-438-2
UN: 3077
ADR: 9 III
Merck Index: 14,8530

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR