6-TAMRA free acid amine-reactive fluorescent dye

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 6-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-4-carboxybenzoate

90008
90008.EA 220 EUR
90008.
6-TAMRA free acid amine-reactive fluorescent dye
6-TAMRA

Free acid form of 6-TAMRA (6-carboxytetramethylrhodamine) single isomer. 6-Carboxytetramethylrhodamine, triethylammonium salt is a single isomer rhodamine dye. The 6-carboxy group can be activated to succinimidyl ester or mixed anhydride for coupling with amines using standard methods.


  • λEx/λEm (MeOH) = 540/565 nm
  • Dark red solid soluble in DMSO, DMF, MeOH, or pH>6.5 buffers
  • Store at ≤ 4 °C and protect from light, especially in solution
  • Isomer purity ≥98%

Formule: C₂₅H₂₂N₂O₅
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01073638
CAS nummer: 91809-67-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30bdce420ac2242855e54660d93a5a1fe401f1c2&vid=30bdce420ac2242855e54660d93a5a1fe401f1c2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR