tert-Butylmethylether 99,0 GC, corrected for water content, BAKER ANALYZED® HPLC HPLC/UHPLC, Spectrophotometry, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether

9042.1000EA 175.46 EUR
9042.1000 9042.2500 9042.5000 9042-03
tert-Butylmethylether 99,0 GC, corrected for water content, BAKER ANALYZED® HPLC HPLC/UHPLC, Spectrophotometry, J.T.Baker®
tert-Butylmethylether

BAKER ANALYZED™ HPLC Reagent—High purity solvents for use in analytical and preparative separations. All are optimized for all of your Liquid Chromatography applications, including HPLC, UHPLC, gel permeation chromatography and UV-spectrophotometric analysis.

Formule: (CH₃)₃COCH₃
MW: 88.15 g/mol
Kookpunt: 55.2
Smeltpunt: –108.6 °C
Dichtheid: 0.7405 g/cm³ (20 °C)
Vlampunt: –28 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008812
CAS nummer: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3 II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 99.0 %
Assay (C₅H₁₂O) (by GC, corrected for water) ≥ 99.0 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤ 0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤ 0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤ 0.1
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤ 215
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤ 0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤ 1.0 ppb
Peroxide (as H₂O₂) ≤ 1 ppm
Residue after Evaporation ≤ 3.0 ppm
Water (by KF, coulometric) ≤ 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR