Fraction collectors, Foxy R1 and R2
Leverancier: COLLECTION PREFIX HPLC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AS-68-3870SA AS-68-3870OL
905-0533EA 0 EUR
905-0533 905-0119
Fraction collectors, Foxy R1 and R2
Fractieverzamelaars
Foxy R1 and Foxy R2 fraction collectors are ideal for a lot of applications including HPLC, preparative HPLC, protein and peptide purification. The Foxy R1 uses a single collection rack for a small footprint. The two-rack Foxy R2 uses radio frequency identification to automatically detect collection racks for easy set-up. Multiple fraction collectors can be 'daisy chained' together to increase fraction capacity.
  • Sturdy cast aluminium frame
  • Symbolic touch screen interface
  • Built-in Ethernet and RS232 communication capabilities
  • Faster tube changes
  • µl delay
More Product Information