Supelco® Organic Reference Standard, Demethoxycurcumin

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

90593-10MG
90593-10MGEA 249 EUR
90593-10MG
Supelco® Organic Reference Standard, Demethoxycurcumin
Standaarden Organische standaarden

  • GHS signal: GHS09

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d66c84093feb26341043eb560bd503647d450b6b&vid=d66c84093feb26341043eb560bd503647d450b6b&prid=web&clid=SAPDAM

Order Now

Specificaties

Synonym(s) (E,E)-1-(4-Hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione; Curcumin II; Desmethoxycurcumin; Monodemethoxycurcumin
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 22608-11-3
MDL No. MFCD03427310
Moleculair gewicht 338,35
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR