HPLC-standaarden, LiChroTest®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.19158.0001 1.19159.0001 1.19157.0001 1.19161.0001 1.19162.0001 1.19163.0001
909-0004EA 290 EUR
909-0004 909-0005 909-0002 909-0007 909-0008 909-0009
HPLC-standaarden, LiChroTest®, Supelco®
Standaarden Standaarden voor chromatografie
Standaardmonsters voor HPLC-systeemkwalificatie.
Merck Millipore en Avantor hebben de LiChroTest® producten ontwikkeld om deze instrumentkwalificatie uit te kunnen voeren in het HPLC-laboratorium. Hiermee bespaart u tijd bij de operationele en prestatiekwalificaties die regelmatig volgens gestandaardiseerde methodes dienen te worden uitgevoerd.
The quality of analytical results is principally determined by the correct functioning of the analytical instruments employed. For this reason, the various quality assurance systems as well as the FDA require analytical instruments to be subjected to periodic qualification. Therefore, before starting a series of analyses, you first should establish whether your HPLC system meets with your requirements. These 'Operational Qualification' (OQ) and 'Performance Qualification' (PQ) steps involve tests of the different modules on their specifications and a check on the entire system using a real application relevant to the laboratory-specific requirements.
Order Now
Specificaties
Environmentally Preferable
M-Clarity quality level MQ100