Methylene blue solution

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

911490ZAEA 59.9 EUR
911490ZA 911490ZAMP
Methylene blue solution
Methylene blue
Formule: C₁₆H₁₈ClN₃S
MDL Nummer: MFCD00150008
CAS nummer: 61-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR