Sudan red G, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 1-[(2-Methoxyphenyl)azo]-2-naphthol, Solvent Red 1, Oil Red 113

M-Clarity™ quality level = MQ100

91282-25MG
91282-25MGEA 140 EUR
91282-25MG
Sudan red G, Supelco®
Sudan red G
Formule: C₁₇H₁₄N₂O₂
Smeltpunt: 225 °C
MDL Nummer: MFCD00046377
CAS nummer: 1229-55-6
EINECS: 214-968-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cf2ade38b7a23099ba463ee5b23fac13606ff36&vid=53a72f08ebd2309ca730389c0bf0cce2708f9232&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR