Supelco® TraceCERT® Organic Reference Standard, Dibenz[a,h]anthracene
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
91861-10MG
91861-10MGEA 82.2 EUR
91861-10MG
Supelco® TraceCERT® Organic Reference Standard, Dibenz[a,h]anthracene
Standaarden Organische standaarden
  • GHS signal: GHS08, GHS09
Certificaten: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Opgelet: P201 - P273 - P308 + P313
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29932b8e7b22c704495f0b9c8b53ce8b1669069a&vid=29932b8e7b22c704495f0b9c8b53ce8b1669069a&prid=web&clid=SAPDAM