Biotin picolyl azide
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
92186
92186.EA 509 EUR
92186.
Biotin picolyl azide
Koppelings- en labelingssystemen voor eiwitten
Biotin picolyl azide reacts with alkyne to form 1,2,3-triazole by 1,3-dipolar Huisgen cycloaddition through the use of a much lower copper (I) concentration without sacrificing reaction efficiency.