Supelco® Organic Reference Standard, D-(+)-Galacturonic acid monohydrate

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

92478-25MG
92478-25MGEA 35.8 EUR
92478-25MG
Supelco® Organic Reference Standard, D-(+)-Galacturonic acid monohydrate
Standaarden Organische standaarden

Order Now

Specificaties

Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 91510-62-2
MDL No. MFCD00071585
Moleculair gewicht 212,15
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR